Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UQCRH, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant UQCRH.
Supplier: Abnova Corporation H00007388A01
Description
Sequence: MGLEDEQKMLTESGDPEEEEEEEEELVDPLTTVREQCEQLEKCVKARERLELCDERVSSRSHTEEDCTEELFDFLHARDHCVAHKLFNNLKSpecifications
UQCRH | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant UQCRH. | |
UQCRH | |
MGC111572/QCR6 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC015177 | |
UQCRH | |
UQCRH (AAH15177, 1 a.a. ∼ 91 a.a) full-length recombinant protein with GST tag. | |
RUO | |
7388 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction