Learn More
Description
Specifications
Specifications
| Antigen | Urotensin II Receptor |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.05% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). |
| Gene Symbols | UTS2R; UR-II-R; UTR; UTR2; GPR14 |
| Host Species | Rabbit |
| Immunogen | Highly purified peptide corresponding to residues 154-167 of rat UTIIR (Accession P49684). |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.