Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133677
Description
USP10 Polyclonal specifically detects USP10 in Human samples. It is validated for Western Blot.Specifications
| USP10 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| USP10 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human USP10. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9100 | |
| Human | |
| IgG |
| Western Blot | |
| 0.27mg/mL | |
| Western Blot 1:500-1:3000 | |
| Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO | |
| Rabbit | |
| 87 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Update cross-reactivity based on sequence identity: Xenopus laevis (93%), Bovine (98%), Xenopus tropicalis (93%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction