Learn More
ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase) (A01), Mouse anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant USP14.
Supplier: Abnova Corporation H00009097A01
Description
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. Mice with a mutation that results in reduced expression of the ortholog of this protein are retarded for growth, develop severe tremors by 2 to 3 weeks of age followed by hindlimb paralysis and death by 6 to 10 weeks of age. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: PQKEVKYEPFSFADDIGSNNCGYYDLQAVLTHQGRSSSSGHYVSWVKRKQDEWIKFDDDKVSIVTPEDILRLSGGGDWHIAYVLLYGPRRVEIMEEESEQSpecifications
ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant USP14. | |
USP14 | |
TGT | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human, Mouse, Rat | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_005151 | |
USP14 | |
USP14 (NP_005142, 395 a.a. ∼ 494 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
9097 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.