Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ubiquitin specific peptidase 18 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human USP18 protein.
Supplier: Abnova Corporation H00011274B01
Description
USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.[supplied by OMIM
Sequence: MSKAFGLLRQICQSILAESSQSPADLEEKKEEDSNMKREQPRERPRAWDYPHGLVGLHNIGQTCCLNSLIQVFVMNVDFTRILKRITVPRGADEQRRSVPFQMLLLLEKMQDSRQKAVRPLELAYCLQKCNVPLFVQHDAAQLYLKLWNLIKDQITDVHLVERLQALYTIRVKDSLICVDCAMESSRNSSMLTLPLSLFDVDSKPLKTLEDALHCFFQPRELSSKSKCFCENCGKKTRGKQVLKLTHLPQTLTIHLMRFSIRNSQTRKICHSLYFPQSLDFSQILPMKRESCDAEEQSGGQYELFAVIAHVGMADSGHYCVYIRNAVDGKWLCFNDSNICLVSWEDIQCTYGNPNYHWQETAYLLVYMKMECSpecifications
ubiquitin specific peptidase 18 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human USP18 protein. | |
USP18 | |
ISG43/UBP43 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC014896 | |
USP18 | |
USP18 (AAH14896, 1 a.a. ∼ 372 a.a) full-length human protein. | |
RUO | |
Yes | |
11274 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction