Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600893
Description
USP18 Polyclonal specifically detects USP18 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
USP18 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
EC 3.1.2.15, EC 3.4.19.-, hUBP43, ISG15-specific-processing protease, ISG43UBP43, ubiquitin specific peptidase 18, ubiquitin specific protease 18, ubl carboxyl-terminal hydrolase 18, ubl thioesterase 18,43 kDa ISG15-specific protease, Ubl thiolesterase 18 | |
Rabbit | |
43.01 kDa | |
0.1 mg | |
Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway | |
11274 | |
Human, Orangutan | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
0.9 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:10000-1:50000, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
Q9UMW8 | |
USP18 | |
This affinity purified USP18 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 25-50 of human USP18 protein. (Uniprot: Q9UMW8) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity-purified antibody is directed against human USP18 protein. A BLAST analysis was used to suggest reactivity with this protein from human, chimpanzee and orangutan based on 100% homology for the immunogen sequence. The core 7 amino acid sequence QPRERPR of the 10 amino acid immunogen sequence may be reactive with USP18 from mouse and dog. This core sequence also shows partial homology with proteins other than USP18. Cross reactivity with USP18 homologues from other sources has not been determined. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction