Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP21 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512024
Description
This antibody is predicted to react with bovine based on sequence homology.
USP21 is a ubiquitin-specific protease, an enzyme that removes ubiquitin from ubiquitinated proteins. The encoded protein belongs to the C19 peptidase family, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. This protein has been reported to be capable of removing NEDD8 from NEDD8 conjugates.
Specifications
USP21 | |
Polyclonal | |
Unconjugated | |
Q9UK80 | |
USP21 | |
KLH conjugated synthetic peptide between 406-436 amino acids from the C-terminal region of human USP21 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
USP21 | |
Ubiquitin carboxyl-terminal hydrolase 21; Ubiquitin thioesterase 21; Ubiquitin-specific-processing protease 21; Deubiquitinating enzyme 21; NEDD8-specific protease; USP21; USP23; | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
27005 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction