Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ubiquitin specific peptidase 45, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant USP45.
Supplier: Abnova Corporation H00085015A01
Description
Sequence: FKSSRTEPHCIIINLSTWIIWCYECDEKLSTHCNKKVLAQIVDFLQKHASKTQTSAFSRIMKLCEEKCETDEIQKGGKCRNLSVRGITNLGSpecifications
ubiquitin specific peptidase 45 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant USP45. | |
USP45 | |
MGC14793 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
XM_371838 | |
USP45 | |
USP45 (XP_371838, 106 a.a. ∼ 196 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
85015 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction