Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP7 Mouse anti-Human, Mouse, Clone: 1G7, MilliporeSigma™

Catalog No. 051946MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-194-6MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-194-6MI Supplier MilliporeSigma Supplier No. 051946
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-USP7 Antibody, clone 1G7 is validated for use in western blot, immunoprecipitation, immunohistochemistry for the detection of USP7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP7
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1G7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003461.2
Gene Symbols USP7, TEF1, HAUSP
Host Species Mouse
Immunogen Histadine-tagged recombinant protein corresponding to human USP7.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.