Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512004
Description
This antibody is predicted to react with mouse based on sequence homology.
USP8 is a ubiquitin specific protease that plays an important regulatory role at the level of protein turnover by preventing degradation. USP8 is involved in cell proliferation, and probably regulates the stability of STAM2 and RASGRF1. USP8 may regulate T-cell anergy mediated by RNF128 via the formation of a complex containing RNF128 and STAM2. As revealed by structur/function studies, USP8 forms a ternary complex with RNF128 and OTUB1, and interacts with the SH3 domain of STAM2 and RASGRF1. Expression of USP8 is induced Upon growth stimulation in starved human fibroblasts, and expression decreases in response to growth arrest induced by cell-cell contact.
Specifications
USP8 | |
Polyclonal | |
USP8 | |
Ubiquitin carboxyl-terminal hydrolase 8, Ubiquitin thiolesterase 8, Ubiquitin-specific processing protease 8, Deubiquitinating enzyme 8, hUBPy | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
9101 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P40818 | |
USP8 | |
KLH conjugated synthetic peptide between 1058-1087 amino acids from the C-terminal region of human USP8 (UBPY) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction