Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

utrophin, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-852 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-852 Supplier Abnova Corporation Supplier No. H00007402A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant UTRN.

This gene shares both structural and functional similarities with the dystrophin gene. It contains an actin-binding N-terminus, a triple coiled-coil repeat central region, and a C-terminus that consists of protein-protein interaction motifs which interact with dystroglycan protein components. The protein encoded by this gene is located at the neuromuscular synapse and myotendinous junctions, where it participates in post-synaptic membrane maintenance and acetylcholine receptor clustering. Mouse studies suggest that this gene may serve as a functional substitute for the dystrophin gene and therefore, may serve as a potential therapeutic alternative to muscular dystrophy which is caused by mutations in the dystrophin gene. Alternative splicing of the utrophin gene has been described; however, the full-length nature of these variants has not yet been determined. [provided by RefSeq

Sequence: LEARMQILEDHNKQLESQLHRLRQLLEQPESDSRINGVSPWASPQHSALSYSLDPDASGPQFHQAAGEDLLAPPHDTSTDLTEVMEQIHSTFPSCCPNVPSRPQAM

Specifications

Antigen utrophin
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UTRN.
Formulation 50% glycerol
Gene UTRN
Gene Accession No. NM_007124
Gene Alias DMDL/DRP/DRP1/FLJ23678
Gene Symbols UTRN
Host Species Mouse
Immunogen UTRN (NP_009055, 3328 a.a. ∼ 3433 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7402
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.