Learn More
Vα 11.1, 11.2[b,d] TCR Rat anti-Mouse, FITC, Clone: RR8-1, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553222
Description
The RR8-1 antibody reacts with the Vα 11.1 and Vα 11.2, but not Vα 11.3 T-cell Receptors (TCR) of mice having the b (e.g., C57BL) and d (e.g., DBA/1, DBA/2, NZW) haplotypes of Tcra gene complex. RR8-1 antibody does not react with strains
Host Species: Rat
Clone: RR8-1
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse T-Cell Clone B10
Flow Cytometry

Specifications
Vα 11.1, 11.2[b,d] TCR | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse T-Cell Clone B10 | |
0.25 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
RR8-1 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.