Learn More
Vβ 11 TCR Rat anti-Mouse, PE, Clone: RR3-15, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553198
Description
The RR3-15 antibody reacts with the Vβ 11 T-Cell Receptor (TCR) of mice having the b haplotype (e.g., A, C57BL, C58, DBA/1) of the Tcrb gene complex. The Tcrb-V11 gene locus is deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) and c (e.g., R
Host Species: Rat
Clone: RR3-15
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Cytolytic T-Cell Clone OH6
Flow Cytometry

Specifications
Vβ 11 TCR | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Cytolytic T-Cell Clone OH6 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
RR3-15 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.