Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ 17[a] T Mouse anti-Mouse, FITC, Clone: KJ23, BD

Catalog No. BDB553212 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553212 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553212 Supplier BD Biosciences Supplier No. 553212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KJ23 antibody reacts with the Vβ 17[a] T-cell Receptor (TCR) of mice having the a haplotype (eg,C57L, SJL, SWR) of the Tcrb gene complex. Strains having the b (eg, A, AKR, BALB/c, CBA,C3H/He, C57BL, C58, DBA/1, DBA/2) Tcrb haplotype do not expres

Host Species: Mouse
Clone: KJ23
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse T cells

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 17[a] T
Applications Flow Cytometry
Classification Monoclonal
Clone KJ23
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse T cells
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.