Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vγ 2 TCR Hamster anti-Mouse, FITC, Clone: UC3-10A6, BD

Catalog No. BDB553226 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553226 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553226 Supplier BD Biosciences Supplier No. 553226

Hamster Monoclonal Antibody

The UC3-10A6 antibody reacts with Vγ 2 T-cell Receptor (TCR)-bearing T lymphocytes, which make up significant proportions of γδ TCR-bearing cells in the late fetal and adult thymus and adult peripheral lymphoid tissues and lung. The freq

Host Species: Hamster
Clone: UC3-10A6
Isotype: IgG1 κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vγ 2 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone UC3-10A6
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Not Reported
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.