Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vα 3.2[b,c] TCR Rat anti-Mouse, FITC, Clone: RR3-16, BD

Catalog No. BDB553219 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553219 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553219 Supplier BD Biosciences Supplier No. 553219
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RR3-16 antibody reacts with the Vα 3.2 T-Cell Receptor (TCR) of mice having the b (e.g., C57BL) and c (e.g., SWR, SJL, NZB, NOD) haplotypes of the Tcra gene complex, but not with TCR encoded by other members of Tcra-V3 gene subfamily. RR3-16 ant

Host Species: Rat
Clone: RR3-16
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Cytolytic T-Cell Clone OH6

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vα 3.2[b,c] TCR
Applications Flow Cytometry
Classification Monoclonal
Clone RR3-16
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Cytolytic T-Cell Clone OH6
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.