Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vγ 3 TCR Hamster anti-Mouse, FITC, Clone: 536, BD

Catalog No. BDB553229 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553229 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553229 Supplier BD Biosciences Supplier No. 553229
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 536 antibody reacts with Vγ 3† T-cell Receptor (TCR)-bearing T lymphocytes, which are the predominant γδ TCRbearing cells in the early fetal thymus and the adult epidermis of euthymic mice. The first T cells to mature in the e

Host Species: Hamster
Clone: 536
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: AKR mouse dendritic epidermal cell clone 7-17

Flow Cytometry, Immunofluorescence

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vγ 3 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone 536
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen AKR mouse dendritic epidermal cell clone 7-17
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.