Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vδ 4 TCR Hamster anti-Mouse, FITC, Clone: GL2, BD

Catalog No. BDB552143 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552143 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552143 Supplier BD Biosciences Supplier No. 552143
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The GL2 antibody reacts with Vδ 4 T-cell Receptor (TCR)-bearing T cells, which are the predominant γδ TCR-bearing cells in the adult intestinal epithelium (γδ IEL). The frequency of Vδ 4 TCR-bearing γδ IEL differs among inbred strains of mice and may be influenced by haplotypes of the MHC, Tcrg, and/or Tcrd gene complexes. There is evidence that these γδ IEL develop in the absence of thymic influence. T lymphocytes expressing Vδ 4 TCR have also been found in lactating mammary glands, the spleen, and the thymus.

Host Species: Hamster
Clone: GL2
Isotype: IgG2 κ
Species Reactivity: Mouse
Immunogen: G57BL/6 mouse intestinal intra-epithelial lymphocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vδ 4 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone GL2
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen G57BL/6 mouse intestinal intra-epithelial lymphocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.