Learn More
Vβ 4 TCR Rat anti-Mouse, FITC, Clone: KT4, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553365
Description
The KT4 antibody reacts with the Vβ4 T-cell Receptor (TCR) of mice having the a (e.g., C57BR, C57L, SJL, SWR), b (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2), and c (e.g., RIII) haplotypes of the Tcrb gene complex. Vβ4 TCR-beari
Host Species: Rat
Clone: KT4
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: KT4 B10.D2 mouse T-cell clone I3
Flow Cytometry

Specifications
Vβ 4 TCR | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
KT4 B10.D2 mouse T-cell clone I3 | |
0.25 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
KT4 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.