Learn More
Vβ 5.1, 5.2 TCR Mouse anti-Mouse, PE, Clone: MR9-4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 553190
Description
The MR9-4 antibody reacts with the Vβ 5.1 and Vβ 5.2 T-cell Receptors of strains having the b haplotype (e.g., C57BL) of the Tcrb gene complex. These gene loci are deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII) Tcrb
Host Species: Mouse
Clone: MR9-4
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse T-Cell Hybridoma 2HB51.8
Flow Cytometry

Specifications
Vβ 5.1, 5.2 TCR | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse T-Cell Hybridoma 2HB51.8 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
MR9-4 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.