Learn More
Vβ 6 TCR Rat anti-Mouse, Biotin, Clone: RR4-7, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553192
Description
The RR4-7 antibody reacts with the Vβ 6 T-Cell Receptor (TCR) of mice having the a (e.g., C57BR, C57L, SJL) and b (e.g., A, BALB/c, CBA/Ca, C3H/He, C57BL, DBA/1) haplotypes of the Tcrb gene complex. The Tcrb-V6 gene locus is deleted in mice having th
Host Species: Rat
Clone: RR4-7
Isotype: IgG2b λ
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Helper T-Cell Clone OI11
Flow Cytometry

Specifications
Vβ 6 TCR | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
C57BL/6 Mouse Helper T-Cell Clone OI11 | |
0.25 mg | |
Immunology | |
Mouse | |
IgG2b λ |
Flow Cytometry | |
RR4-7 | |
Biotin | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.