Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ 7 TCR Rat anti-Mouse, FITC, Clone: TR310, BD

Catalog No. BDB553215 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553215 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553215 Supplier BD Biosciences Supplier No. 553215

Rat Monoclonal Antibody

The TR310 antibody reacts with the Vβ 7 T-Cell Receptor (TCR) of strains having the a (e.g., C57BR, C57L, SJL), b (e.g., A, BALB/c, CBA/Ca, C3H/He, C57BL, DBA/1), and c (e.g., RIII) haplotypes of the Tcrb gene complex. Vβ 7 is among the predomin

Host Species: Rat
Clone: TR310
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: AKR/Cum Mouse spontaneous T Lymphoma Cell Line TK1

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 7 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone TR310
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen AKR/Cum Mouse spontaneous T Lymphoma Cell Line TK1
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.