Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ 8.3 T-Cell Receptor Hamster anti-Mouse, PE, Clone: 1B3.3, BD

Catalog No. BDB553664 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553664 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553664 Supplier BD Biosciences Supplier No. 553664
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 1B3.3 antibody reacts with the Vβ 8.3 T-cell Receptor (TCR), but not the Vβ 8.1 nor Vβ 8.2 TCR, of mice having the b haplotype (e.g., A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. The Tcrb-V8 subfamily gene loci are deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII) haplotype. Vβ 8.3 is one of the restricted TCR β chains expressed by NK-T cells. Staphylococcal enterotoxin B, in association with antigen-presenting cells expressing I-A and/or I-E, stimulates lymphocytes bearing Vβ 8 TCR and selectively eliminates those T cells in vivo. Vβ 8.3 TCR-bearing T-cell clones can be activated by 1B3.3 mAb. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.

Host Species: Hamster
Clone: 1B3.3
Isotype: IgG3, λ1
Species Reactivity: Mouse
Immunogen: Vβ 8.3 TCR-expressing mouse Th1 clone 3.L2

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 8.3 T-Cell Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone 1B3.3
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Vβ 8.3 TCR-expressing mouse Th1 clone 3.L2
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG3
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.