Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ8 Mouse anti-Human, Unlabeled, Clone: JR2, BD

Catalog No. BDB555604 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555604 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555604 Supplier BD Biosciences Supplier No. 555604
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the β-chain variable region (Vβ8) of the T-cell antigen receptor expressed on 0.5% to 3% of peripheral blood T cells. This antibody has been further characterized as having specificity to a major subset of the BV8 region as determined in the TCR Workshop of July 1995. Staphylococcal enterotoxin E has been shown to specifically stimulate T cells bearing Vβ8 (BV8). This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ8
Applications Flow Cytometry
Classification Monoclonal
Clone JR2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description BV8
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.