Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vγ9 TCR Mouse anti-Human, FITC, Clone: B3, BD

Catalog No. BDB555732 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555732 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555732 Supplier BD Biosciences Supplier No. 555732
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the human variable γ9 subunit (Vγ9) of γ/δ T-cell receptor (TCR). The γ/δ TCR is composed of two disulfide-linked glycoproteins. The gamma chain is approximately 45-60kDa and is detected on a small subset peripheral blood T cells and thymocytes. The majority of normal peripheral blood γ/δ T cells express a Vγ9+/Vδ2+ phenotype. The reason for this selection in the T-cell repertoire is not well understood.

Host Species: Mouse
Clone: B3
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vγ9 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone B3
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.