Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

v-Src Mouse anti-Mouse, Clone: 327, MilliporeSigma™

Catalog No. MABS193MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS193MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS193MI Supplier MilliporeSigma Supplier No. MABS193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-v-Src Antibody, clone 327 detects level of v-Src and has been published and validated for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen v-Src
Applications Western Blot
Classification Monoclonal
Clone 327
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005408
Gene Symbols SRC, SRC1
Host Species Mouse
Immunogen Recombinant protein corresponding to purified pp60src from bacterial recombinants which direct the synthesis of v-Src.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.