Learn More
VAC14, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human VAC14 protein.
Supplier: Abnova Corporation H00055697B01P
Description
Phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2) is a low-abundance signaling molecule. A regulatory complex made up of VAC14 and FIG4 (MIM 609390) control synthesis of PI(3,5)P2 by activating PI(3)P kinase, FAB1 (PIP5K3; MIM 609414). The VAC14/FIG4 complex also functions in the breakdown of PI(3,5)P2 (Zhang et al., 2007 [PubMed 17956977]).[supplied by OMIM
Sequence: MNPEKDFAPLTPNIVRALNDKLYEKRKVAALEIEKLVREFVAQNNTVQIKHVIQTLSQEFALSQHPHSRKGGLIGLAACSIALGKDSGLYLKELIEPVLTCFNDADSRLRYYACEALYNIVKVARGAVLPHFNVLFDGLSKLAADPDPNVKSGSELLDRLLKDIVTESNKFDLVSFIPLLRERIYSNNQYARQFIISWILVLESVPDINLLDYLPEILDGLFQILGDNGKEIRKMCEVVLGEFLKEIKKNPSSVKFAEMANILVIHCQTTDDLIQLTAMCWMREFIQLAGRVMLPYSSGILTAVLPCLAYDDRKKSIKEVANVCNQSLMKLVTPEDDELDELRPGQRQAEPTPDDALPKQEGTASGGPDGSCDSSFSSGISVFTAASTERAPVTLHLDGIVQVLNCHLSDTAIGMMTRIAVLKWLYHLYIKTPRKMFRHTDSLFPILLQTLSDESDEVILKDLEVLAEIASSPAGQTDDPGPLDGPDLQASHSELQVPTPGRAGLLNTSGTKGLECSPSTPTMNSYFYKFMINLLKRFSSERKLLEVRGPFIIRQLCLLLNAENIFHSMADILLREEDLKFASTMVHALNTILLTSTELFQLRNQLKDLKTLESQNLFCCLYRSWCHNPVTTVSLCFLTQNYRHAYDLIQKFGDLEVTVDFLAEVDKLVQLIECPIFTYLRLQLLDVKNNPYLIKALYGLLMLLPQSSAFQLLSHRLQCVPNPELLQTEDSLKAAPKSQKADSPSIDYAELLQHFEKVQNKHLEVRHQRSGRGDHLDRRVVLSpecifications
VAC14 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human VAC14 protein. | |
VAC14 | |
ArPIKfyve/FLJ10305/FLJ36622/FLJ46582/MGC149815/MGC149816/TAX1BP2/TRX | |
Mouse | |
Affinity chromatography | |
RUO | |
55697 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_018052.3 | |
VAC14 | |
VAC14 (NP_060522.3, 1 a.a. ∼ 782 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.