Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vesicle-associated membrane protein 8 (endobrevin), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016951 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-951 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-951 Supplier Abnova Corporation Supplier No. H00008673B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human VAMP8 protein.

Synaptobrevins/VAMPs, syntaxins, and the 25kDa synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. It is associated with the perinuclear vesicular structures of the early endocytic compartment. It has been found that VAMP8 interacts specifically with the soluble NSF-attachment protein (alpha-SNAP), most likely through an VAMP8-containing SNARE complex. [provided by RefSeq

Sequence: MEEASEGGGNDRVRNLQSEVEGVKNIMTQNVERILARGENLEHLRNKTEDLEATSEHFKTTSQKVARKFWWKNVKMIVLICVIVFIIILFIVLFATGAFS

Specifications

Antigen vesicle-associated membrane protein 8 (endobrevin)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human VAMP8 protein.
Formulation PBS with no preservative; pH 7.4
Gene VAMP8
Gene Accession No. BC001634
Gene Alias EDB
Gene Symbols VAMP8
Host Species Mouse
Immunogen VAMP8 (AAH01634, 1 a.a. ∼ 100 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8673
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.