Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid Receptor Like Protein 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5398
Description
Anti-Vanilloid Receptor Like Protein 1 Antibody, pain, C-terminus detects level of Vanilloid Receptor Like Protein 1 and has been published and validated for use in immunohistochemistry.
Specifications
Vanilloid Receptor Like Protein 1 | |
Polyclonal | |
Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50μL of sterile distilled water. Centrifuge if necessary to remove any residue. | |
Rabbit | |
50 μL | |
NM_016113.3 | |
Lyophilized: −20-−70°C for up to 6 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Immunohistochemistry | |
Unconjugated | |
TRPV2; VRL; TrpV2; VRL-1; OTRPC2; MGC12549; VRL1 | |
Peptide corresponding to the C-terminal 18 amino acids of the rat VRL-1 protein sequence (Acc#: AAD26364). This immunogen sequence is 100% conserved in the mouse growth factor regulated calcium channel (Acc#: AB021665) | |
Primary | |
Rat | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction