Learn More
Varicella-Zoster Virus Mouse anti-Human, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8616
Description
This Anti-Varicella-Zoster Virus Antibody, Immediate Early Gene 62 is validated for use in immunofluoresence, immunoprecipitation, western blot, immunohistochemistry for the detection of Varicella-Zoster Virus.
Specifications
Varicella-Zoster Virus | |
Monoclonal | |
Purified immunoglobulin. In 0.02M PB with 0.25M NaCl, pH=7.6 with 0.1% Sodium azide. | |
VZV HZ strain. | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG1 |
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
100 μg | |
Human | |