Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

valyl-tRNA synthetase, Mouse, Clone: 4B7, Abnova™

Catalog No. 89014211 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-211 Supplier Abnova Corporation Supplier No. H00007407M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant VARS.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. The protein encoded by this gene belongs to class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex. [provided by RefSeq

Sequence: AVRLSNQGFQAYDFPAVTTAQYSFWLYELCDVYLECLKPVLNGVDQVAAECARQTLYTCLDVGLRLLSPFMPFVTEELFQRLPRRMPQAPPSLCVTPYPEPSECSWKDP

Specifications

Antigen valyl-tRNA synthetase
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant VARS.
Formulation PBS with no preservative; pH 7.4
Gene VARS
Gene Accession No. NM_006295
Gene Alias G7A/VARS1/VARS2
Gene Symbols VARS
Host Species Mouse
Immunogen VARS (NP_006286, 994 a.a. ∼ 1102 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7407
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.