Learn More
VASA/DDX4 Human, Unlabeled, Clone: R11-A3, BD
Human Monoclonal Antibody
Supplier: BD Biosciences 560189
Description
The R11-A3 Fab reacts with a protein of the Asp-Glu-Ala-Asp (DEAD) box (DDX) family that is homologous to the VASA protein of Drosophila. The VASA/DDX4 gene is conserved in invertebrate and vertebrate species, and its expression is germ cell specific. In the human, VASA/DDX4 is expressed in fetal and adult germ cells, both male and female, but not in other fetal and adult tissues. Studies of VASA/DDX4 expression in various types of tumors suggest that expression of VASA/DDX4 is a useful biomarker for germ cell cancers.
The R11-A3 Fab was selected by screening a human Fab phage display library (Hoet et al.) licensed from Dyax Corp (Cambridge, MA) with immobilized recombinant human VASA/DDX4 protein.
Host Species: Human
Clone: R11-A3
Isotype: Ig, λ
Species Reactivity: Mouse
Immunohistochemistry, Western Blotting

Specifications
VASA/DDX4 | |
Monoclonal | |
0.5mg/mL | |
DDX4 | |
Human | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Western Blot | |
R11-A3 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
immunogen is NA for phage display technology | |
0.1 mg | |
Cell Biology | |
Human, Mouse | |
Ig λ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.