Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vasopressin Rabbit anti-Human, Mouse, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™

Catalog No. AB1565MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1565MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1565MI Supplier MilliporeSigma Supplier No. AB1565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Vasopressin Antibody detects level of Vasopressin and has been published and validated for use in immunocytochemistry, immunohistochemistry (paraffin), RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vasopressin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat rabbit antisera. Liquid, with 0.1% sodium azide.
Gene Symbols AVP; AVP-NPII; VP; AVRP; ARVP; ADH
Host Species Rabbit
Immunogen Arginine vasopressin (Sigma) conjugated to thyroglobulin.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000490.3
Target Species Human, Mouse, Rabbit, Rat, Sheep
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.