Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vasopressin V1B Receptor. Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB9311MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9311MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9311MI Supplier MilliporeSigma Supplier No. AB9311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Vasopressin V1B Receptor. Antibody is validated for use in immunohistochemistry (paraffin) for the detection of Vasopressin V1B Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vasopressin V1B Receptor.
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS, pH 7.7 with 0.01% sodium azide.
Gene Symbols V1BR; AVPR1B; AVPR3; VPR3
Host Species Rabbit
Immunogen Synthetic peptide from the 3rd cytoplasmic domain of human Vasopressin V1B Receptor.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.