Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VASP Mouse, Unlabeled, Clone: 43, BD

Catalog No. BDB610448 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610448 150 μg
BDB610447 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610448 Supplier BD Biosciences Supplier No. 610448
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vasodilator-stimulated phosphoprotein (VASP), a substrate for cAMP- and cGMP-dependent kinases, is associated with actin filaments, focal adhesions, and dynamic membrane regions. VASP is composed of several distinct domains: a central L-proline-rich domain contains a GPPPPP motif as a single copy and as a three-fold tandem repeat, as well as three conserved phosphorylation sites for cyclic nucleotide-dependent protein kinases (Ser157, Ser239, and Thr278). A C-terminal domain contains a repetitive mixed-charge cluster which is predicted to form an α-helix. The C-terminal domain appears to be responsible for anchoring at focal adhesion sites. VASP has been shown to be a ligand for profilins. Profilins bind to the poly-L-proline motifs of VASP and it is postulated that these two molecules act in concert to convey signal transduction to actin filament formation.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen VASP
Applications Western Blot
Classification Monoclonal
Clone 43
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.