Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vav 1 guanine nucleotide exchange factor, Mouse, Clone: 1A6, Abnova™

Catalog No. 89001046 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-046 Supplier Abnova Corporation Supplier No. H00007409M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant VAV1.

The protein encoded by this proto-oncogene is a member of the Dbl family of guanine nucleotide exchange factors (GEF) for the Rho family of GTP binding proteins. The protein is important in hematopoiesis, playing a role in T-cell and B-cell development and activation. This particular GEF has been identified as the specific binding partner of Nef proteins from HIV-1. Coexpression and binding of these partners initiates profound morphological changes, cytoskeletal rearrangements and the JNK/SAPK signaling cascade, leading to increased levels of viral transcription and replication. [provided by RefSeq

Sequence: RGLTELVEFYQQNSLKDCFKSLDTTLQFPFKEPEKRTISRPAVGSTKYFGTAKARYDFCARDRSELSLKEGDIIKILNKKGQQGWWRGEIYGRVGWFPANYVEEDYSEYC

Specifications

Antigen vav 1 guanine nucleotide exchange factor
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant VAV1.
Formulation PBS with no preservative; pH 7.4
Gene VAV1
Gene Accession No. BC013361
Gene Alias VAV
Gene Symbols VAV1
Host Species Mouse
Immunogen VAV1 (AAH13361, 681 a.a. ∼ 790 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7409
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.