Learn More
ventral anterior homeobox 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human VAX2 protein.
Supplier: Abnova Corporation H00025806B01
Description
This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development. [provided by RefSeq
Sequence: MGDGGAERDRGPARRAESGGGGGRCGDRSGAGDLRADGGGHSPTEVAGTSASSPAGSRESGADSDGQPGPGEADHCRRILVRDAKGTIREIVLPKGLDLDRPKRTRTSFTAEQLYRLEMEFQRCQYVVGRERTELARQLNLSETQVKVWFQNRRTKQKKDQSRDLEKRASSSASEAFATSNILRLLEQGRLLSVPRAPSLLALTPSLPGLPASHRGTSLGDPRNSSPRLNPLSSASASPPLPPPLPAVCFSSAPLLDLPAGYELGSSAFEPYSWLERKVGSASSCKKANTSpecifications
ventral anterior homeobox 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human VAX2 protein. | |
VAX2 | |
DRES93 | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_012476.1 | |
VAX2 | |
VAX2 (NP_036608.1, 1 a.a. ∼ 290 a.a) full-length human protein. | |
RUO | |
Yes | |
25806 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.