Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vb 13 TCR Mouse anti-Mouse, PE, Clone: MR12-3, BD

Catalog No. BDB561541 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561541 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561541 Supplier BD Biosciences Supplier No. 561541

Mouse Monoclonal Antibody

The MR12-3 monoclonal antibody specifically binds to the Vβ 13 T-cell Receptor (TCR) of mice having the b haplotype (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. The Tcrb-V13 c (e.g., RIII) haplotype.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vb 13 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone MR12-3
Concentration 0.2mg/mL
Conjugate PE
Description TCR V beta 13; TCR Vβ 13; TCR Vβ13; Vβ 13 T-cell Receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Not reported
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.