Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-VCAM-1/CD106, PerCP, Clone: IE10, R&D Systems™
Supplier: R&D Systems FAB5649C
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| VCAM1 | |
| Activated HUVEC human umbilical vein endothelial cells | |
| 100 Tests | |
| Primary | |
| The antibody was screened using COS cells transfected with cDNAs for E-Selectin, VCAM-1, ICAM-1 and PECAM-1 and was shown to be specific for human VCAM-1/CD106. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| IE10 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7412 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction