Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM-1 Mouse anti-Human, Clone: P8B1, MilliporeSigma™

Catalog No. MAB2511MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2511MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2511MI Supplier MilliporeSigma Supplier No. MAB2511
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-VCAM-1 Antibody, clone P8B1 detects level of VCAM-1 and has been published and validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VCAM-1
Applications Functional Assay
Classification Monoclonal
Clone P8B1
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.2Mphosphate buffer to 0.25MNaCl, pH 7.6 with 0.1% sodium azide. Caution: Sodium azide is toxic. See online MSDS.
Gene Accession No. NM_001078.2; NM_080682.1
Gene Symbols VCAM1, INCAM-100, MGC99561, CD106, DKFZp779G2333, L1CAM
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.