Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-VCP Clone: 4A8,

Catalog No. ST1553100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1553100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1553100UG Supplier MilliporeSigma™ Supplier No. ST1553100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects VCP Clone: 4A8 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting

VCP is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VCP
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4A8
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant human VCP (aa 1-589), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽67kDa VCP protein in HePG2 cells and human placenta tissue.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.