Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vd2 TCR Mouse anti-Human, FITC, Clone: B6, BD

Catalog No. BDB562088 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562088 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562088 Supplier BD Biosciences Supplier No. 562088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the human variable δ 2 subunit (V δ 2) of γ/ δ T cell receptor (TCR). The γ/ δ TCR is composed of two disulfide linked glycoproteins. The delta chain is approximately 40-60 kDa and is restricted to peripheral blood T cells and thymocytes. The majority of normal peripheral blood γδ 2[+] phenotype. The reason for this selection in the T-cell repertoire is not well understood.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vd2 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone B6
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.