Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VDP p115 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217245
Description
VDP p115 Polyclonal specifically detects VDP p115 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VDP p115 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| USO1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human VDP/p115. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 8615 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| p115, Transcytosis-associated protein, USO1 homolog, vesicle docking protein (yeast), USO1 vesicle docking protein homolog (yeast), vesicle docking protein, vesicle docking protein p115 | |
| Rabbit | |
| 108 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (93%), cow (97%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction