Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-cadherin Mouse anti-Human, Clone: BV6, MilliporeSigma™

Catalog No. MABT134MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT134MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT134MI Supplier MilliporeSigma Supplier No. MABT134
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-VE-cadherin Antibody, clone BV6 is validated for use in western blot, immunocytochemistry, flow cytometry, immunohistochemistry (paraffin) for the detection of VE-cadherin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VE-cadherin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone BV6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001786
Gene Symbols CDH5, CD144, 7B4
Host Species Mouse
Immunogen HUVEC cells
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.