Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071420MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-142-0MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-142-0MI Supplier MilliporeSigma Supplier No. 071420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-VEGF Antibody detects level of VEGF and has been published and validated for use in western blot, immunohistochemistry (paraffin), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified rabbit polyclonal IgG employing immobilized mouse VEGF matrix. Antibody solution (1mg/mL) was lyophilized from PBS, pH 7.2. Reconstitute with sterile distilled water.
Gene Symbols VEGFA; VPF; VEGF-A; VEGF
Host Species Rabbit
Immunogen Highly pure (>98%) recombinant mouse Vascular Endothelial Growth Factor.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001025366.1
Target Species Human, Mouse
Content And Storage 2-8°C for up to 6 weeks, −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.