Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Mouse anti-Human, Rat, Clone: CH-10, MilliporeSigma™

Catalog No. 051116MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-111-6MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-111-6MI Supplier MilliporeSigma Supplier No. 051116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-VEGF Antibody, clone CH-10 is an antibody against VEGF for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Western Blot
Classification Monoclonal
Clone CH-10
Conjugate Unconjugated
Formulation Liquid in 20mM sodium phosphate, 250mM NaCl, pH. 7.6 containing 0.1% sodium azide.
Gene Accession No. NP_033531.3
Gene Symbols MGC70609, MVCD1, VEGF, VEGF-A, VPF
Host Species Mouse
Immunogen Recombinant mouse VEGF.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.