Learn More
VEGF Mouse anti-Human, Unlabeled, Clone: G143-850, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 554539
Description
The G143-850 antibody recognizes vascular endothelial growth factor (VEGF) by western blotting. VEGF is a homodimeric heavily glycosylated protein. Human VEGF occurs in several molecular variants which arise by alternative splicing of the mRNA. Splice forms of VEGF differ in biological properties such as the receptor types which they recognize. Various forms of VEGF can be detected at MW 18-25kDa in SDS-PAGE.
Host Species: Mouse
Clone: G143-850
Isotype: IgG2a
Species Reactivity: Human
Immunoprecipitation, Western Blotting

Specifications
VEGF | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Western Blot | |
G143-850 | |
Unconjugated | |
Mouse | |
0.5 mg | |
Immunology | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.