Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Mouse anti-Human, Unlabeled, Clone: G153-694, BD

Catalog No. BDB555036 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555036 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555036 Supplier BD Biosciences Supplier No. 555036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vascular endothelial growth factor (VEGF) is a heparin-binding, dimeric protein related to the PDGF/sis family of growth factors. Major sources of VEGF include pituitary cells, monocytes/macrophages, smooth muscle, and keratinocytes. VEGF is a mitogen for endothelial cells, activates and is chemoattractant for monocytes, enhances blood vessel permeability, and is a pro-coagulant. Human VEGF occurs in several molecular variants arising by alternative splicing of the mRNA. The splice forms of VEGF differ in biological properties. VEGF is a homodimeric heavily glycosylated protein of 46-48kDa (18-25kDa subunits). Glycosylation is not required, however, for biological activity. The subunits are linked by disulphide bonds. Different isoforms of VEGF have different properties in vitro and this may apply also to their in vivo functions.

Immunofluorescence, Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Immunohistochemistry
Classification Monoclonal
Clone G153-694
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.