Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Mouse anti-Human, Mouse, Rat, Clone: VG1 , MilliporeSigma™

Catalog No. 051117MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-111-7MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-111-7MI Supplier MilliporeSigma Supplier No. 051117
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect VEGF using this Anti-VEGF Antibody, clone VG1 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Western Blot
Classification Monoclonal
Clone VG1
Conjugate Unconjugated
Formulation Unpurified mouse monoclonal IgG1κ in ascites fluid containing no preservatives.
Gene Accession No. NP_033531.3
Gene Symbols MGC70609, VPF, VEGF-A, VEGF, MVCD1
Host Species Mouse
Immunogen Recombinant human VEGF 189aa isoform
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.