Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Receptor-1 Mouse anti-Human, Mouse, MilliporeSigma™

Catalog No. MAB1664MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1664MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1664MI Supplier MilliporeSigma Supplier No. MAB1664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-VEGF Receptor-1 Antibody is an antibody against VEGF Receptor-1 for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF Receptor-1
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified from Ascites. Liquid in PBS and 5% Trehalose.
Gene Accession No. NM_002019.3
Gene Symbols FLT1, VEGFR1, FLT, FRT, Flt-1, VEGFR-1, EC 2.7.10.1
Host Species Mouse
Immunogen Human recombinant Flt-1 (extracellular domain). Actual epitope recognized by this clone is between 1-250 aa of human Flt-1.
Quantity 50 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.